Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pdcl2
Entrez Gene ID
498352
Gene Name
phosducin-like 2
Gene Aliases
RGD1559712
Chromosome Location
Chr.14: 34384125 - 34404920 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_573585.6 XP_573585.3 3 - 4 557 94

View Full Product Details