The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106201.1 | NP_001099671.1 | 8 - 9 | 3049 | 112 |
XM_006231634.1 | XP_006231696.1 | 9 - 10 | 3725 | 112 | |
XM_006231635.2 | XP_006231697.1 | 9 - 10 | 3663 | 112 |