Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nolc1
Entrez Gene ID
64896
Gene Name
nucleolar and coiled-body phosphoprotein 1
Gene Aliases
Nopp140
Chromosome Location
Chr.1: 265829057 - 265839819 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022869.1 NP_074060.1 3 - 4 373 68
XM_006231560.2 XP_006231622.1 3 - 4 424 68
GenBank mRNA BC126088.1 - 3 - 4 338 68
M94287.1 - 3 - 4 373 68

View Full Product Details