The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021689.1 | NP_067721.1 | 2 - 3 | 271 | 111 |
GenBank mRNA | AF074952.1 | - | 2 - 3 | 271 | 111 |