The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012036.1 | NP_001012036.1 | 1 - 1 | 1496 | 115 |
GenBank mRNA | BC079277.1 | - | 1 - 1 | 1496 | 115 |