Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccnb1ip1
Entrez Gene ID
498492
Gene Name
cyclin B1 interacting protein 1, E3 ubiquitin protein ligase
Gene Aliases
-
Chromosome Location
Chr.15: 27720220 - 27736308 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025141.1 NP_001020312.1 2 - 3 255 111
XM_006251901.2 XP_006251963.1 3 - 4 665 111
XM_006251903.2 XP_006251965.1 2 - 3 768 111
XM_008770563.1 XP_008768785.1 2 - 3 769 111
XM_008770564.1 XP_008768786.1 3 - 4 666 111
XM_008770565.1 XP_008768787.1 3 - 4 539 111
XM_008770566.1 XP_008768788.1 2 - 3 769 111
XM_008770567.1 XP_008768789.1 2 - 3 768 111
XM_008770568.1 XP_008768790.1 2 - 3 768 111
GenBank mRNA BC091423.1 - 2 - 3 255 111

View Full Product Details