Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Otx2
Entrez Gene ID
305858
Gene Name
orthodenticle homeobox 2
Gene Aliases
-
Chromosome Location
Chr.15: 25500037 - 25510602 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100566.1 NP_001094036.1 - 165 101
GenBank mRNA CB580080.1 - - 165 101

Gene Symbol
LOC100911492
Entrez Gene ID
100911492
Gene Name
homeobox protein OTX2-like
Gene Aliases
-
Chromosome Location
Chr.15: 25516697 - 25526560 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006251836.2 XP_006251898.1 2 - 3 444 101
XM_008770615.1 XP_008768837.1 2 - 3 444 101

View Full Product Details