Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ero1l
Entrez Gene ID
171562
Gene Name
ERO1-like (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.15: 19619947 - 19655381 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA FQ228432.1 - - 1934 168

View Full Product Details