Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cd84
Entrez Gene ID
501872
Gene Name
CD84 molecule
Gene Aliases
RGD1564553
Chromosome Location
Chr.13: 90241321 - 90278392 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008769753.1 XP_008767975.1 2 - 3 477 92
XM_008769754.1 XP_008767976.1 3 - 4 544 92
XM_008769755.1 XP_008767977.1 2 - 3 473 92
XM_008769756.1 XP_008767978.1 2 - 3 475 92

View Full Product Details