Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adck3
Entrez Gene ID
360887
Gene Name
aarF domain containing kinase 3
Gene Aliases
Cabc1
Chromosome Location
Chr.13: 98451629 - 98480438 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006250387.1 XP_006250449.1 1 - 2 111 56

View Full Product Details