Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ovol2
Entrez Gene ID
296201
Gene Name
ovo-like zinc finger 2
Gene Aliases
Zfp339
Chromosome Location
Chr.3: 138433990 - 138463690 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006235124.2 XP_006235186.1 1 - 2 1226 126

View Full Product Details