The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108592.2 | NP_001102062.1 | 6 - 7 | 571 | 73 |
XM_006235116.2 | XP_006235178.1 | 6 - 7 | 732 | 73 | |
GenBank mRNA | BC168760.1 | - | 6 - 7 | 571 | 73 |
FQ213679.1 | - | 6 - 7 | 603 | 73 |