Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slco4a1
Entrez Gene ID
171144
Gene Name
solute carrier organic anion transporter family, member 4a1
Gene Aliases
OATP-E, Slc21a12
Chromosome Location
Chr.3: 175838994 - 175857985 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133608.2 NP_598292.2 6 - 7 1447 121
GenBank mRNA AF239262.1 - 5 - 6 1281 121
BC097285.1 - 6 - 7 1447 121
BC097286.1 - 6 - 7 1389 121

View Full Product Details