Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mtg2
Entrez Gene ID
296462
Gene Name
mitochondrial ribosome-associated GTPase 2
Gene Aliases
Gtpbp5
Chromosome Location
Chr.3: 175448313 - 175464109 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013924.1 NP_001013946.1 2 - 3 208 61
XM_006235820.2 XP_006235882.1 2 - 3 262 61
XM_008762493.1 XP_008760715.1 2 - 3 285 61
GenBank mRNA BC083707.1 - 2 - 3 208 61

View Full Product Details