The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191913.1 | NP_001178842.1 | 6 - 7 | 749 | 66 |
XM_006256881.2 | XP_006256943.1 | 7 - 8 | 1090 | 66 | |
XM_006256883.2 | XP_006256945.1 | 7 - 8 | 931 | 66 |