The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007756.1 | NP_001007757.1 | 3 - 4 | 579 | 105 |
GenBank mRNA | BC079160.1 | - | 3 - 4 | 579 | 105 |