The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127531.2 | NP_001121003.1 | 4 - 5 | 553 | 94 |
GenBank mRNA | BC166480.1 | - | 4 - 5 | 553 | 94 |
FQ235170.1 | - | 1 - 2 | 48 | 94 |