Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Glis1
Entrez Gene ID
298732
Gene Name
GLIS family zinc finger 1
Gene Aliases
-
Chromosome Location
Chr.5: 127043344 - 127233395 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006238561.2 XP_006238623.1 4 - 5 1443 67

View Full Product Details