Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc17
Entrez Gene ID
500528
Gene Name
coiled-coil domain containing 17
Gene Aliases
-
Chromosome Location
Chr.5: 135441995 - 135445794 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014067.2 NP_001014089.2 11 - 12 1770 78
XM_006238702.2 XP_006238764.1 10 - 11 2000 78
XM_006238703.2 XP_006238765.1 11 - 12 1934 78
XM_006238704.2 XP_006238766.1 11 - 12 1928 78
XM_006238706.2 XP_006238768.1 6 - 7 1544 78
XM_006238707.2 XP_006238769.1 12 - 13 1862 78
GenBank mRNA AA926261.1 - 1 - 2 78
BC087059.1 - 9 - 10 2014 78

View Full Product Details