The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039010.2 | NP_001034099.2 | 2 - 3 | 610 | 68 |
XM_008763998.1 | XP_008762220.1 | 2 - 3 | 395 | 68 | |
XM_008763999.1 | XP_008762221.1 | 2 - 3 | 180 | 68 | |
GenBank mRNA | CK474267.1 | - | 2 - 3 | 649 | 68 |