Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdc20
Entrez Gene ID
64515
Gene Name
cell division cycle 20
Gene Aliases
p55cdc
Chromosome Location
Chr.5: 137260728 - 137264931 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171993.1 NP_741990.1 3 - 4 616 123
GenBank mRNA BC085691.1 - 3 - 4 600 123
U05341.1 - 3 - 4 616 123

View Full Product Details