Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tal1
Entrez Gene ID
313507
Gene Name
T-cell acute lymphocytic leukemia 1
Gene Aliases
Tal-1
Chromosome Location
Chr.5: 133864403 - 133878604 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008763967.1 XP_008762189.1 1 - 2 654 71

View Full Product Details