The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009710.1 | NP_001009710.1 | 5 - 6 | 1777 | 79 |
XM_006238682.2 | XP_006238744.1 | 5 - 6 | 2162 | 79 | |
GenBank mRNA | BC087037.1 | - | 5 - 6 | 1777 | 79 |