Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Txndc12
Entrez Gene ID
298370
Gene Name
thioredoxin domain containing 12 (endoplasmic reticulum)
Gene Aliases
RGD1305960
Chromosome Location
Chr.5: 128450830 - 128475858 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100840.1 NP_001094310.1 1 - 2 232 86
GenBank mRNA BC100255.1 - 1 - 2 174 86
BC158737.1 - 1 - 2 178 86
CV117277.1 - 1 - 2 469 86
FQ218354.1 - 1 - 2 129 86

View Full Product Details