The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100840.1 | NP_001094310.1 | 6 - 7 | 587 | 70 |
GenBank mRNA | BC100255.1 | - | 6 - 7 | 529 | 70 |
BC158737.1 | - | 6 - 7 | 533 | 70 | |
FQ218354.1 | - | 6 - 7 | 484 | 70 |