Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem59
Entrez Gene ID
100196907
Gene Name
transmembrane protein 59
Gene Aliases
RGD1310313
Chromosome Location
Chr.5: 126783061 - 126804177 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001139465.2 NP_001132937.1 3 - 4 444 70
GenBank mRNA CK476149.1 - 3 - 4 449 70
FM094601.1 - 2 - 3 176 70
FQ212504.1 - 3 - 4 410 70
FQ214069.1 - 3 - 4 441 70
FQ226828.1 - 2 - 3 164 70
FQ231829.1 - 3 - 4 428 70
FQ232106.1 - 3 - 4 449 70

View Full Product Details