The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001044275.1 | NP_001037740.1 | 7 - 8 | 776 | 69 |
GenBank mRNA | BC085795.1 | - | 7 - 8 | 776 | 69 |