Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ssbp3
Entrez Gene ID
84354
Gene Name
single stranded DNA binding protein 3
Gene Aliases
Ssdp, Ssdp3
Chromosome Location
Chr.5: 126508099 - 126649830 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053358.1 NP_445810.1 12 - 13 1122 90
XM_006238532.2 XP_006238594.1 13 - 14 1304 90
XM_006238533.2 XP_006238595.1 13 - 14 1279 90
XM_006238534.2 XP_006238596.1 13 - 14 1261 90
XM_006238535.2 XP_006238597.1 13 - 14 1235 90
XM_006238536.2 XP_006238598.1 12 - 13 1221 90
XM_006238537.2 XP_006238599.1 12 - 13 1195 90
XM_006238538.2 XP_006238600.1 12 - 13 1177 90
XM_006238539.2 XP_006238601.1 10 - 11 1161 90
XM_008763902.1 XP_008762124.1 13 - 14 2957 90
XM_008763903.1 XP_008762125.1 14 - 15 2511 90
XM_008763904.1 XP_008762126.1 13 - 14 2930 90
XM_008763905.1 XP_008762127.1 13 - 14 2912 90
XM_008763906.1 XP_008762128.1 13 - 14 2885 90
XM_008763907.1 XP_008762129.1 12 - 13 2872 90
XM_008763908.1 XP_008762130.1 12 - 13 2846 90
XM_008763909.1 XP_008762131.1 12 - 13 2801 90
GenBank mRNA AF121893.1 - 12 - 13 1122 90
AY724490.1 - 13 - 14 903 90
BC107666.1 - 12 - 13 1106 90
FQ230168.1 - 7 - 8 400 90

View Full Product Details