Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ssbp3
Entrez Gene ID
84354
Gene Name
single stranded DNA binding protein 3
Gene Aliases
Ssdp, Ssdp3
Chromosome Location
Chr.5: 126508099 - 126649830 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006238532.2 XP_006238594.1 8 - 9 951 64
XM_006238533.2 XP_006238595.1 8 - 9 950 64
XM_006238534.2 XP_006238596.1 8 - 9 950 64
XM_006238535.2 XP_006238597.1 8 - 9 948 64
XM_006238536.2 XP_006238598.1 7 - 8 868 64
XM_006238537.2 XP_006238599.1 7 - 8 866 64
XM_006238538.2 XP_006238600.1 7 - 8 866 64
XM_006238539.2 XP_006238601.1 5 - 6 808 64
XM_008763902.1 XP_008762124.1 8 - 9 2604 64
XM_008763903.1 XP_008762125.1 9 - 10 2158 64
XM_008763904.1 XP_008762126.1 8 - 9 2601 64
XM_008763905.1 XP_008762127.1 8 - 9 2601 64
XM_008763906.1 XP_008762128.1 8 - 9 2598 64
XM_008763907.1 XP_008762129.1 7 - 8 2519 64
XM_008763908.1 XP_008762130.1 7 - 8 2517 64
XM_008763909.1 XP_008762131.1 7 - 8 2514 64
GenBank mRNA AY724490.1 - 8 - 9 616 64
BC107666.1 - 7 - 8 819 64
FQ230168.1 - 2 - 3 63

View Full Product Details