Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Orc1
Entrez Gene ID
313479
Gene Name
origin recognition complex, subunit 1
Gene Aliases
Orc1l
Chromosome Location
Chr.5: 128186617 - 128210795 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177931.2 NP_808792.1 1 - 2 94 93
XM_006238515.2 XP_006238577.1 2 - 3 296 93
XM_006238516.2 XP_006238578.1 3 - 4 409 93
XM_006238518.2 XP_006238580.1 2 - 3 296 93
GenBank mRNA AB105378.1 - 1 - 2 94 93

View Full Product Details