Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Foxe3
Entrez Gene ID
171302
Gene Name
forkhead box E3
Gene Aliases
FREAC-8, HFH7, HNF3
Chromosome Location
Chr.5: 133724796 - 133725656 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134339.1 NP_599166.1 1 - 1 52 97

View Full Product Details