Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
B4galt2
Entrez Gene ID
313536
Gene Name
UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 2
Gene Aliases
-
Chromosome Location
Chr.5: 136707510 - 136717346 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107965.1 NP_001101435.1 1 - 2 137 65

View Full Product Details