Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmprss11a
Entrez Gene ID
686581
Gene Name
transmembrane protease, serine 11A
Gene Aliases
-
Chromosome Location
Chr.14: 23226735 - 23284515 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008770103.1 XP_008768325.1 6 - 7 555 111

View Full Product Details