The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013109.1 | NP_001013127.1 | 7 - 8 | 667 | 80 |
XM_006243306.2 | XP_006243368.1 | 11 - 12 | 1184 | 80 | |
GenBank mRNA | BC091410.1 | - | 7 - 8 | 667 | 80 |