The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025123.3 | NP_001020294.3 | 7 - 8 | 1405 | 77 |
GenBank mRNA | BC093594.1 | - | 7 - 8 | 1405 | 77 |
BC100617.1 | - | 7 - 8 | 1404 | 77 | |
FQ211817.1 | - | 6 - 7 | 1109 | 77 |