The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012540.2 | NP_036672.2 | 5 - 6 | 1256 | 83 |
XM_006243150.2 | XP_006243212.1 | 5 - 6 | 1253 | 83 | |
GenBank mRNA | X00469.1 | - | 5 - 6 | 1256 | 83 |
X17160.1 | - | - | 1266 | 83 |