Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Btg4
Entrez Gene ID
315650
Gene Name
B-cell translocation gene 4
Gene Aliases
SCIR-27
Chromosome Location
Chr.8: 55492781 - 55508100 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013176.1 NP_001013194.1 1 - 2 172 80
GenBank mRNA BC088755.1 - 1 - 2 172 80

View Full Product Details