Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmprss4
Entrez Gene ID
367074
Gene Name
transmembrane protease, serine 4
Gene Aliases
-
Chromosome Location
Chr.8: 49469804 - 49503306 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108998.1 NP_001102468.1 12 - 13 1556 67
XM_006242949.2 XP_006243011.1 13 - 14 2338 67

View Full Product Details