Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lctl
Entrez Gene ID
315750
Gene Name
lactase-like
Gene Aliases
-
Chromosome Location
Chr.8: 69073915 - 69090405 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108158.1 NP_001101628.1 6 - 7 783 153

View Full Product Details