Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Stoml1
Entrez Gene ID
300748
Gene Name
stomatin (EPB72)-like 1
Gene Aliases
RGD1559463
Chromosome Location
Chr.8: 63037156 - 63045024 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006243184.2 XP_006243246.1 5 - 6 925 64
XM_236297.8 XP_236297.3 6 - 7 1075 64

View Full Product Details