Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Drd2
Entrez Gene ID
24318
Gene Name
dopamine receptor D2
Gene Aliases
-
Chromosome Location
Chr.8: 53678777 - 53743643 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012547.1 NP_036679.1 3 - 4 742 89
XM_006242979.2 XP_006243041.1 3 - 4 523 89
XM_008766190.1 XP_008764412.1 2 - 3 759 89
XM_008766191.1 XP_008764413.1 2 - 3 759 89
GenBank mRNA M36831.1 - 3 - 4 523 89
X53278.1 - 3 - 4 522 89
X56065.1 - 3 - 4 742 89

View Full Product Details