Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr61
Entrez Gene ID
363064
Gene Name
WD repeat domain 61
Gene Aliases
RGD1308228
Chromosome Location
Chr.8: 59296339 - 59313626 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006243085.2 XP_006243147.1 1 - 2 69 112

View Full Product Details