Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lman1l
Entrez Gene ID
300743
Gene Name
lectin, mannose-binding, 1 like
Gene Aliases
slamp
Chromosome Location
Chr.8: 62386806 - 62399648 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012465.1 NP_001012483.1 2 - 3 357 89
GenBank mRNA AB188302.1 - 2 - 3 357 89

View Full Product Details