Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ddx6
Entrez Gene ID
500988
Gene Name
DEAD (Asp-Glu-Ala-Asp) box helicase 6
Gene Aliases
RGD1564560
Chromosome Location
Chr.8: 48924768 - 48961394 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109292.1 NP_001102762.1 3 - 4 623 146
XM_006242950.2 XP_006243012.1 3 - 4 623 146
XM_006242951.2 XP_006243013.1 3 - 4 637 146
XM_006242952.2 XP_006243014.1 3 - 4 632 146
XM_006242954.2 XP_006243016.1 3 - 4 906 146
GenBank mRNA FQ213450.1 - 3 - 4 608 146

View Full Product Details