Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc37a4
Entrez Gene ID
29573
Gene Name
solute carrier family 37 (glucose-6-phosphate transporter), member 4
Gene Aliases
G6pt1
Chromosome Location
Chr.8: 48716939 - 48723024 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031589.2 NP_113777.2 7 - 8 1267 65
XM_008766153.1 XP_008764375.1 6 - 7 1028 65
GenBank mRNA BC070889.1 - 7 - 8 1267 65

View Full Product Details