Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bcl9l
Entrez Gene ID
300673
Gene Name
B-cell CLL/lymphoma 9-like
Gene Aliases
-
Chromosome Location
Chr.8: 48805684 - 48835794 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106817.1 NP_001100287.1 3 - 4 822 75
XM_006242923.2 XP_006242985.1 3 - 4 2384 75
XM_006242924.2 XP_006242986.1 2 - 3 2327 75

View Full Product Details