Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nono
Entrez Gene ID
317259
Gene Name
non-POU domain containing, octamer-binding
Gene Aliases
-
Chromosome Location
Chr.X: 71324365 - 71342225 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012356.1 NP_001012356.1 10 - 11 1316 80
GenBank mRNA BC089880.1 - 10 - 11 1316 80

View Full Product Details