Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pdzd11
Entrez Gene ID
302422
Gene Name
PDZ domain containing 11
Gene Aliases
RGD1560007
Chromosome Location
Chr.X: 70458594 - 70461635 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106945.1 NP_001100415.1 3 - 4 366 70
XM_006257068.2 XP_006257130.1 3 - 4 272 70

View Full Product Details