The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126373.2 | NP_001119845.2 | 10 - 11 | 1449 | 86 |
GenBank mRNA | BC162023.1 | - | 10 - 11 | 1442 | 86 |