Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Map7d2
Entrez Gene ID
317508
Gene Name
MAP7 domain containing 2
Gene Aliases
Brelin, Mtap7d2, RGD1564852
Chromosome Location
Chr.X: 38400578 - 38467825 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289778.1 NP_001276707.1 7 - 8 958 109
GenBank mRNA AB266744.1 - 7 - 8 958 109

View Full Product Details